ENA423200254 Enamine

(1R)-1-(5-chloropyridin-2-yl)ethan-1-ol

  • MDL number MFCD23855690
  • Cas Number : 1567887-32-4
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-1-(5-chloropyridin-2-yl)ethan-1-ol
Inchi Code 1S/C7H8ClNO/c1-5(10)7-3-2-6(8)4-9-7/h2-5,10H,1H3/t5-/m1/s1
Inchi Key OLWSYXWXQLPXCG-RXMQYKEDSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-157665

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-157665
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart