ENA423200256 Enamine

4-ethoxy-1H-indole

  • MDL number MFCD11110381
  • Cas Number : 23456-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123787
Availability details for
Quantity
  • Availability
  • Add to Cart