ENA423200262 Enamine

(1-amino-3-ethoxy-2,2-dimethylcyclobutyl)methanol

  • MDL number MFCD27980675
  • Cas Number : 1602002-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-157719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157719
Availability details for
Quantity
  • Availability
  • Add to Cart