ENA423200268 Enamine

2-chloro-6-(3-chloro-4-fluorophenoxy)benzaldehyde

  • MDL number MFCD19536730
  • Cas Number : 1484597-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157733

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157733
Availability details for
Quantity
  • Availability
  • Add to Cart