ENA423200279 Enamine

[2-methyl-4-(trifluoromethyl)phenyl]methanamine

  • MDL number MFCD20724401
  • Cas Number : 874571-73-0
  • Molecular Weight : 189.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-methyl-4-(trifluoromethyl)phenyl]methanamine
Inchi Code 1S/C9H10F3N/c1-6-4-8(9(10,11)12)3-2-7(6)5-13/h2-4H,5,13H2,1H3
Inchi Key VMJKHBXPGXRDHD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-157755

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-157755
pictograms GHS05,GHS07
hstatements H227,H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart