ENA423200283 Enamine

7-oxa-4-azaspiro[2.5]octan-5-one

  • MDL number MFCD11655976
  • Cas Number : 1100753-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157762
Availability details for
Quantity
  • Availability
  • Add to Cart