ENA423200299 Enamine

7-fluoroquinoline

  • MDL number MFCD01685513
  • Cas Number : 396-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124531
Availability details for
Quantity
  • Availability
  • Add to Cart