ENA423200303 Enamine

1-hydroxy-3-methylbutan-2-one

  • MDL number MFCD18968789
  • Cas Number : 36960-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157803
Availability details for
Quantity
  • Availability
  • Add to Cart