ENA423200304 Enamine

2-(4-chlorophenyl)-1,3-benzothiazol-6-amine

  • MDL number MFCD01796944
  • Cas Number : 53544-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157804

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157804
Availability details for
Quantity
  • Availability
  • Add to Cart