ENA423200306 Enamine

1-nitro-3-(trichloromethyl)benzene

  • MDL number MFCD00051691
  • Cas Number : 709-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157806
Availability details for
Quantity
  • Availability
  • Add to Cart