ENA423200307 Enamine

3-methyl-3-phenylpyrrolidin-2-one

  • MDL number MFCD24552174
  • Cas Number : 51439-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-84
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157808
Availability details for
Quantity
  • Availability
  • Add to Cart