ENA423200310 Enamine

2-[3-(morpholin-4-yl)phenyl]acetic acid hydrochloride

  • MDL number MFCD27980683
  • Cas Number : 1607326-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157812

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-157812
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart