ENA423200314 Enamine

7-{[(tert-butoxy)carbonyl]amino}-8,8-dimethyl-2-oxabicyclo[4.2.0]octane-7-carboxylic acid

  • MDL number MFCD27980684
  • Cas Number : 1797019-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157816
Availability details for
Quantity
  • Availability
  • Add to Cart