ENA423200315 Enamine

2-methylpentane-1,5-diol

  • MDL number MFCD00971964
  • Cas Number : 42856-62-2
  • Molecular Weight : 118.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-1,5-pentanediol
Inchi Code 1S/C6H14O2/c1-6(5-8)3-2-4-7/h6-8H,2-5H2,1H3
Inchi Key AAAWJUMVTPNRDT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-157818

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-157818
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart