ENA423200319 Enamine

N-methoxyacetamide

  • MDL number MFCD20760911
  • Cas Number : 5806-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157823

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-157823
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart