ENA423200330 Enamine

2,2,2-trichloro-1-(1-methyl-4-nitro-1H-pyrrol-2-yl)ethan-1-one

  • MDL number MFCD03789117
  • Cas Number : 120122-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157846

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157846
Availability details for
Quantity
  • Availability
  • Add to Cart