ENA423200333 Enamine

(3R)-3-(propan-2-yloxy)pyrrolidine

  • MDL number MFCD11858551
  • Cas Number : 1187932-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157849
Availability details for
Quantity
  • Availability
  • Add to Cart