ENA423200335 Enamine

2-chlorocyclohexane-1-carbonitrile

  • MDL number MFCD20631632
  • Cas Number : 89728-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157852

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157852
Availability details for
Quantity
  • Availability
  • Add to Cart