ENA423200336 Enamine

2-({[(pyridin-2-yl)methyl]amino}methyl)phenol

  • MDL number MFCD08674997
  • Cas Number : 63671-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-157853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157853
Availability details for
Quantity
  • Availability
  • Add to Cart