ENA423200340 Enamine

2-(2-amino-1,2,3,4-tetrahydronaphthalen-2-yl)acetic acid hydrochloride

  • MDL number MFCD27980690
  • Cas Number : 1607266-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157861
Availability details for
Quantity
  • Availability
  • Add to Cart