ENA423200341 Enamine

2-methyl-3-(phenylsulfanyl)aniline

  • MDL number MFCD17285192
  • Cas Number : 1344215-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157862
Availability details for
Quantity
  • Availability
  • Add to Cart