ENA423200342 Enamine

2-(5-chloro-1,2,3,4-tetrahydroisoquinolin-2-yl)ethan-1-amine

  • MDL number MFCD26668376
  • Cas Number : 1369275-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-157868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157868
Availability details for
Quantity
  • Availability
  • Add to Cart