ENA423200346 Enamine

4-(2,2-dimethylmorpholin-4-yl)-2-ethoxyaniline

  • MDL number MFCD18605498
  • Cas Number : 1405012-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157878
Availability details for
Quantity
  • Availability
  • Add to Cart