ENA423200354 Enamine

4-bromo-2,5-dimethoxybenzene-1-sulfonamide

  • MDL number MFCD17925818
  • Cas Number : 1206129-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157890
Availability details for
Quantity
  • Availability
  • Add to Cart