ENA423200368 Enamine

1-(aminomethyl)-N,N-dimethylcyclobutan-1-amine

  • MDL number MFCD17170813
  • Cas Number : 176445-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157934

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157934
Availability details for
Quantity
  • Availability
  • Add to Cart