ENA423200393 Enamine

1-(3-cyanophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12583969
  • Cas Number : 1153039-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131045
Availability details for
Quantity
  • Availability
  • Add to Cart