ENA423200397 Enamine

8-iodo-1H,2H,3H,4H,5H-pyrrolo[1,2-a][1,4]diazepin-1-one

  • MDL number MFCD27980704
  • Cas Number : 1416221-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158029
Availability details for
Quantity
  • Availability
  • Add to Cart