ENA423200401 Enamine

1,3-dimethyl-5-(1H-1,2,4-triazol-1-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11191346
  • Cas Number : 1152565-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158067
Availability details for
Quantity
  • Availability
  • Add to Cart