ENA423200404 Enamine

ethyl 4-amino-3-bromo-1H-pyrazole-5-carboxylate

  • MDL number MFCD18632904
  • Cas Number : 1979126-28-7
  • Molecular Weight : 234.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-amino-3-bromo-1H-pyrazole-5-carboxylate
Inchi Code 1S/C6H8BrN3O2/c1-2-12-6(11)4-3(8)5(7)10-9-4/h2,8H2,1H3,(H,9,10)
Inchi Key ILCPMHDLWQWUDN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-158073

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-158073
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart