ENA423200405 Enamine

4-chloropentanenitrile

  • MDL number MFCD21184641
  • Cas Number : 13249-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158075
Availability details for
Quantity
  • Availability
  • Add to Cart