ENA423200407 Enamine

[(6-methoxypyridin-2-yl)methyl](methyl)amine

  • MDL number MFCD13188748
  • Cas Number : 1060806-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-158079
Availability details for
Quantity
  • Availability
  • Add to Cart