ENA423200408 Enamine

ethyl 4-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-3-oxobutanoate

  • MDL number MFCD01926123
  • Cas Number : 13855-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131667
Availability details for
Quantity
  • Availability
  • Add to Cart