ENA423200410 Enamine

3,3-dimethylazepane

  • MDL number MFCD22395547
  • Cas Number : 54152-50-0
  • Molecular Weight : 127.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,3-dimethylazepane
Inchi Code 1S/C8H17N/c1-8(2)5-3-4-6-9-7-8/h9H,3-7H2,1-2H3
Inchi Key MCJWZDIFFOGAJS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-158084

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-158084
pictograms GHS02,GHS05,GHS07
hstatements H226,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart