ENA423200412 Enamine

4-(4-chlorophenyl)-3,3-dimethylbutanoic acid

  • MDL number MFCD19320004
  • Cas Number : 1089303-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-70
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158086
Availability details for
Quantity
  • Availability
  • Add to Cart