ENA423200425 Enamine

(cyclopropylmethyl)[(4-methylmorpholin-2-yl)methyl]amine

  • MDL number MFCD21644815
  • Cas Number : 1483002-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159068
Availability details for
Quantity
  • Availability
  • Add to Cart