ENA423200431 Enamine

3,5,5-trimethylhexanamide

  • MDL number MFCD20544728
  • Cas Number : 90726-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159669

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159669
Availability details for
Quantity
  • Availability
  • Add to Cart