ENA423200435 Enamine

5-(5-methyl-1-benzofuran-2-yl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD16061472
  • Cas Number : 1248595-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-159675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159675
Availability details for
Quantity
  • Availability
  • Add to Cart