ENA423200437 Enamine

1-azabicyclo[3.2.1]octan-4-one

  • MDL number MFCD24689969
  • Cas Number : 17604-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159677
Availability details for
Quantity
  • Availability
  • Add to Cart