ENA423200438 Enamine

2-(4-bromophenyl)-1,1,1-trifluoropropan-2-ol

  • MDL number MFCD16067973
  • Cas Number : 122243-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159678

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-159678
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart