ENA423200439 Enamine

2-(aminomethyl)-N-methyl-N-(propan-2-yl)aniline

  • MDL number MFCD11155587
  • Cas Number : 1020969-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159679
Availability details for
Quantity
  • Availability
  • Add to Cart