ENA423200450 Enamine

2-(aminomethyl)cyclopentan-1-one hydrochloride

  • MDL number MFCD21602354
  • Cas Number : 1360547-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159700

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159700
Availability details for
Quantity
  • Availability
  • Add to Cart