ENA423200451 Enamine

1-(bromomethyl)-2-ethylbenzene

  • MDL number MFCD12024947
  • Cas Number : 57825-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159703

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159703
Availability details for
Quantity
  • Availability
  • Add to Cart