ENA423200452 Enamine

methyl 6-fluoro-1H-indole-2-carboxylate

  • MDL number MFCD04966976
  • Cas Number : 136818-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-167
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-159704

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159704
Availability details for
Quantity
  • Availability
  • Add to Cart