ENA423200458 Enamine

4-(5-bromo-6-chloropyrazin-2-yl)morpholine

  • MDL number MFCD12024891
  • Cas Number : 1378867-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159739
Availability details for
Quantity
  • Availability
  • Add to Cart