ENA423200481 Enamine

1-chloro-3-methyl-2-nitrobenzene

  • MDL number MFCD04117977
  • Cas Number : 5367-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 22...23
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159825

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159825
Availability details for
Quantity
  • Availability
  • Add to Cart