ENA423200485 Enamine

(2S)-4-[(tert-butoxy)carbonyl]morpholine-2-carboxylic acid

  • MDL number MFCD06200662
  • Cas Number : 868689-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-134
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-159830

Safety Information

signalword Warning
pcodes P264,P273,P280,P305+P351+P338,P337+P313,P391,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-159830
pictograms GHS07,GHS09
hstatements H319,H411
Availability details for
Quantity
  • Availability
  • Add to Cart