ENA423200490 Enamine

5-methanesulfonyl-1H-indole

  • MDL number MFCD13190320
  • Cas Number : 152879-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159859

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159859
Availability details for
Quantity
  • Availability
  • Add to Cart