ENA423200498 Enamine

[5-(methylsulfanyl)-1,3,4-thiadiazol-2-yl]methanamine

  • MDL number MFCD24644898
  • Cas Number : 933693-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159871

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159871
Availability details for
Quantity
  • Availability
  • Add to Cart