ENA423200500 Enamine

7-azaspiro[4.5]decane

  • MDL number MFCD03175394
  • Cas Number : 176-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159880
Availability details for
Quantity
  • Availability
  • Add to Cart