ENA423200505 Enamine

{3-[(methylsulfanyl)methyl]phenyl}methanamine

  • MDL number MFCD12779654
  • Cas Number : 869297-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159888
Availability details for
Quantity
  • Availability
  • Add to Cart